Would you like to log in again?
Mobile Mortuary Refrigerator, Mortech®
Supplier: Mortech
1036-R101
29000-040EA 40397.39 USD InStock
29000-040
Mobile Mortuary Refrigerator, Mortech®
Refrigerators
This two-body refrigerator operates at 3°C (37°F) and features a self-contained 1/2hp, air-cooled condensing unit
The unit is insulated with 10.2cm (4") foamed-in-place polyurethane insulation. Four 15.2cm (6") dia. casters allow the unit to be easily moved: two rigid casters and the two swivel casters with locking brakes. A corner bumper surrounds the entire perimeter of the unit. A dial thermometer on the front of the refrigerator monitors interior temperature. The front panel is constructed of 22-gauge Type 304 stainless steel with a No. 4 finish. The rest of the exterior and interior walls and ceilings are constructed of 20-gauge stucco-embossed aluminum. A single door with flush-mount hardware provides access to both body compartments. Each compartment is equipped with a body tray constructed of 18-gauge Type 304 stainless steel with a No. 4 finish. Trays are provided with built-in slots and constructed with continuous welded corners. Each compartment also features a three-piece telescopic sliding carriage frame constructed of extruded aluminum. Carriage frames have nylon rollers for easy sliding
Ordering information: Door may be equipped with a lock; please specify when ordering. Additional electrical configurations are available; contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty