Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Whatman™ Filter Funnel Plates, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1950-114 1950-117 1950-109 1950-109 1950-117 1950-114
80085-992EA 189.28 USD InStock
CA80085-990 80085-992 80085-990 CA80085-992 28200-753 CA28200-753
Whatman™ Filter Funnel Plates, Whatman products (Cytiva)
Funnels Membrane Filter Funnels
Designed for use with Whatman glass microfiber filters
Three-piece construction permits quick filter changing and easy cleaning. Reservoir, base and stem are borosilicate glass with ground contact flanges. Clamping pressure provides a leak-tight seal. Solutions pass through filter and all retained solids are deposited within filter circle. Clamps are plastic-coated for chemical resistance. Replaceable filter support plates of transparent acrylic are suitable for most aqueous solutions and some dilute acids at working temperatures to 65°C maximum. Optional polypropylene plate 28200-753 (9cm) is available.
Ideal for water testing and for biomedical, microbiological and clinical chemistry.
Ordering information: Supplied with acrylic filter support plate and three clamps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty