Would you like to log in again?
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/3, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10300263 10300210 10300214 10300263 10300211 10300212 10300213 10300211 10300214 10300210 10300213 10300212
14224-450PK 73 USD InStock
14224-450 CA76423-780 89233-654 76423-780 CA89233-654 CA14224-450 CA14224-444 14224-444 76423-778 10034-630 CA76423-778 CA11012-658
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.01%), Grade 589/3, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Whatman Grade 589/3 ashless filter paper from Cytiva is particularly useful in capturing very fine particulate samples such as insoluble molecules in fats or oils or supporting ash free dry mass.
  • High purity (<0.01% ash) for ash free dry mass methods
  • Blue ribbon filter
  • Very fine precipitate retention (class 2d acc. to DIN 53 135)
  • Herzberg filtration speed of 750 s*
  • Also available prepleated as Grade 589/3 ½
*The time taken to filter 100 ml water (20° C) through a filter area of 10 cm² at a constant pressure of 5 cm water column.
Grade 589/3 filter paper from Cytiva's business supports accurate measurement of ash free dry mass. The blue ribbon filter is an established component in a variety of quantitative analysis methods, such as quantification of insoluble contaminants in animal and vegetable fats and oils. Ash free dry mass.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty