Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 43, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Whatman™
1443-150 1443-125 1443-185 1443-110 1443-110 1443-150 1443-185 1443-090 1443-125 1443-090
28481-295PK 88.02 USD InStock
CA28481-295 28481-295 70240-046 CA70240-046 28481-302 CA28297-320 28297-320 CA28481-302 70240-050 CA70240-050
Whatman™ Quantitative Filter Papers, Ashless Grades (ash 0.007%), Grade 43, Whatman products (Cytiva)
Filters Filter Paper Quantitative Filter Paper
Cytiva's Whatman Grade 43 filters support inorganic analysis and other forms of instrumental analyses. The low ash content also makes Grade 43 appropriate for gravimetric analyses requiring sample combustion.

  • Very low ash content (0.007% ash concentration maximum)
  • Intermediate particle retention rating of 16 µm
  • Herzberg filtration speed of 155 s
  • Particle collection between that of Grades 40 and 41

Cytiva offers other ashless grades to ensure that you have the appropriate particle retention rating for your needs.

Particle collection and inorganic analysis. Whatman Grade 43 from Cytiva's business can be ashed during gravimetric analysis without contributing substantially to the measured weight.

These filters are also appropriate for:
- Inorganic analysis in the construction, mining, and steel industries
- Particle collection for air pollution monitoring for subsequent analysis by X-Ray fluorescence (XRF) techniques
- Soil analysis
- Foodstuffs filtration analysis

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty