Would you like to log in again?
Whatman™ Grade GF/B Glass Microfiber Filters, Binder Free, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1821-021 1821-914 1821-125 1821-150 1821-024 1821-271 1821-271 1821-185 1821-185 1821-915 1821-025 1821-150 1821-025 1821-042 1821-070 1821-047 1821-055 1821-042 1821-110 1821-024 1821-037 1821-090 1821-070 1821-055 1821-914 1821-090 1821-915
28497-470PK 190.46 USD InStock
CA11013-134 28497-470 28497-492 CA28497-481 10035-298 28497-437 28497-459 28497-454 CA28497-528 CA28497-426 28497-517 CA28497-548 CA11013-942 28497-481 28497-460 CA28497-470 CA28497-492 CA11013-140 28497-404 28497-426 28497-548 10035-300 CA28497-437 CA28497-459 28497-528 28497-506 10035-302
Whatman™ Grade GF/B Glass Microfiber Filters, Binder Free, Whatman products (Cytiva)
Filters Glass Fiber Filters
These borosilicate glass filters are particularly useful for liquid clarification and liquid scintillation counting.
  • Fine particle filter
  • Good loading capacity
  • Binder free
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty