Would you like to log in again?
Corning® Plastic Syringe Filters, Corning
Supplier: Corning
Corning®
431224 431215 431220 431221 431219 431225 431229 431227 431231 431212 431218 431222
28200-040CS 167.47 USD InStock
28200-040 28200-042 28200-030 28200-046 28200-024 28200-034 28200-026 28200-036 28200-014 28200-028 28200-016 28200-018
Corning® Plastic Syringe Filters, Corning
Filters Syringe Filters
Syringe filters are available with a choice of membrane type: Polyethersulfone (PES) for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA), with or without prefilter, for lowest protein binding; Polytetrafluoroethylene (PTFE) for chemical resistance; regenerated cellulose (RC) – the best choice for DMSO compatibility; and nylon (NY) which is hydrophilic, surfactant-free and lowest extractable.
  • 100% integrity tested
  • Certified non pyrogenic and non cytotoxic
Manufactured in accordance with ISO 9002 standards
50 mm filter is recommended as in-line air filter.
Available in 4, 15, 25, 26, and 50 mm diameter filters.Redesigned 26 mm filter increases effective filtration area resulting in faster flow rates and higher throughput. 50 mm fluoropolymer resin vent filters help prevent aggressive solvents from damaging vacuum lines and pumps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty