Would you like to log in again?
Whatman™ Pop-Top and Swin-Lok Plastic Filter Holders, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
420100 420200 420400 420400 420100
28163-045PK 409.06 USD InStock
CA28163-023 CA28163-089 28163-045 28163-089 28163-023
Whatman™ Pop-Top and Swin-Lok Plastic Filter Holders, Whatman products (Cytiva)
Filter Holders
These filter holders are designed for microfiltration and ultra cleaning of small volumes of liquids using positive pressure.
  • Accommodate Nuclepore track etched and cast membranes
  • Syringe compatible
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty