Would you like to log in again?
Whatman™ Syringe Type Membrane Holders, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Whatman™
1980-001 1980-001 1980-002 1980-002
28454-680EA 475.89 USD InStock
CA28454-680 CA28454-670 28454-680 28454-670
Whatman™ Syringe Type Membrane Holders, Whatman products (Cytiva)
Filter Holders
These syringe filter holders are made from stainless steel. The holders with Luer lock fittings connected to a standard syringe offer quick and easy clarification, sterilisation and removal of particulates from small volume samples, typically for HPLC applications.
They contain PTFE gaskets and O-rings which allow the membrane to be autoclaved in place without the filter sticking to the holder. Also available in PP.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty