Would you like to log in again?
Whatman™ Grade 2V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
1202-320 1202-400 1202-500 1202-400 1202-150 1202-270 1202-240 1202-385 1202-185 1202-270 1202-125 1202-500 1202-320 1202-500 1202-185 1202-125 1202-150 1202-240 1202-385
70240-038PK 444.46 USD InStock
70240-038 CA70240-038 CA11013-352 CA10811-038 28465-022 10147-912 28465-066 28465-044 28465-088 28465-102 28465-124 CA28465-044 CA28465-088 CA28465-066 CA28465-022 CA28465-102 CA28465-124 10035-256 CA10147-912
Whatman™ Grade 2V Qualitative Filter Papers, Fluted, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Compared with quadrant-folded filters, Cytiva's Whatman Grade 2V pre-folded filter paper exposes extra surface area, which enables a higher loading capacity and shorter filtration time.
  • Medium flow and nominal particle retention rating of 8 µm
  • Herzberg filtration speed of 240s
  • Fluted (pre-pleated) for easy use with conical filter funnels
  • Can be used for soil analysis or general filtration
Grade 2V pre-folded filter sheets are suitable for extracting samples from soil to support contaminant testing in the presence of weakly acidic solutions. Pre-folded filters for seed and soil testing.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty