Box furnaces are efficient, responsive, and reliable for continuous operation to 1700°C
Long-life molybdenum disilicide heating elements provide fast heat-up, resist thermal shock, and can be subjected to rapid cycling over long periods of time without affecting performance. Furnace chamber is constructed of Moldatherm® high-temperature ceramic fiber insulation. Fan cooling and double shell construction keeps the outer shell cool to the touch. Removable louvered panels provide easy access to the heating elements and Type-B thermocouple.
Ordering information: Furnaces feature a one-year parts and labor warranty. Furnaces supplied without controller and interconnecting wiring. Controllers are 300-segment, 30-program programmable functions. Includes a replaceable hearth plate and RS-485 data connection port.