Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Accessories for Synthware Stopcock, 1:5 PTFE Plug, T-Bore, Kemtech America
Supplier: Kemtech America
S201635
60005-134EA 46.92 USD InStock
60005-134
Accessories for Synthware Stopcock, 1:5 PTFE Plug, T-Bore, Kemtech America
Stopcocks
Replacement Teflon® t-bore stopcock.
  • Teflon® stopcock construction
  • T-bore
  • 16/35 plug size
Comes complete with one Teflon® plug, one Teflon® washer, one O-ring.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty