Would you like to log in again?
Synthware Stopcock, High Vacuum, 1:10 Glass Plug, Kemtech America
Supplier: Kemtech America
Synthware
S332044 S331740 S331230
60005-150EA 65.23 USD InStock
60005-150 60005-154 60005-152
Synthware Stopcock, High Vacuum, 1:10 Glass Plug, Kemtech America
Stopcocks
High vacuum stopcock.
  • Straight bore
  • Heavy wall borosilicate construction
  • All glass stopcock
The 1:10 taper straight bore glass plug is precision ground and hand-lapped with corresponding numbers on the barrel and plug. Stopcocks are supplied with a stainless steel constant pressure retention spring and clip.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty