Would you like to log in again?
Lindberg/Blue M Tube Furnaces, 1700 °C, Thermo Scientific
Supplier: Thermo Scientific
Lindberg/Blue M
STF54434C STF54454C
30784-090EA 23238.15 USD InStock
30784-090 52346-028
Lindberg/Blue M Tube Furnaces, 1700 °C, Thermo Scientific
Ovens Tube Furnaces
Tube furnaces provide excellent temperature uniformity and thermal efficiency throughout the radiant temperature range
Programmable controllers (sold separately) provide 300 programmable segments, 30 programs, and up to 99 segments per program. Self-tuning feature automatically sets the best PID settings for the thermal process. Two LED displays simultaneously indicate actual temperature vs. setpoint. High limit over-temperature control is optional.
Long-life molybdenum disilicide heating elements provide fast heat-up, resist thermal shock, and can be subjected to rapid cycling over long periods of time without affecting performance. Lower shell temperature is achieved using double-shell Moldatherm® construction and fan cooling. Removable louvered panels provide easy access to heating elements and Type-B thermocouple. For horizontal operation only. Standard process tube O.D.: 7.62 cm (3"). Maximum temperature: 1700°C.
Ordering information: Furnaces feature a one-year parts and labor warranty. Supplied with one standard 7.62cm (3") vestibule. Controller and interconnecting power wiring required, not included with furnace. Optional vestibules for 2.54cm (1") or 5.1 (2") process tubes available upon request. Process tubes not included.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty