Would you like to log in again?
Heidolph® Hei-Mix S Magnetic Stirrer
Supplier: Heidolph
Heidolph®
036110460CA 036110460 036110460
89029-824EA 622.27 USD InStock
89029-824 CA11008-218 CA11009-520
Heidolph® Hei-Mix S Magnetic Stirrer
Stirrers Magnetic Stirrers
Magnetic stirrer with white top plate and analogue control dials for speed setting, unit is ideal for titrations.
  • Chemically resistant PVDF top plate and polyamide housing
  • Electronic speed control without digital display
Certifications: IP protection class according to DIN EN 60529: IP 21
Ordering information: For heating baths, support rods, spins bars and other accessories, contact your Avantor representative.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty