Would you like to log in again?
Corning® BioCoat® Poly-D-Lysine/Laminin 8-well Culture Slide, Corning
Supplier: Corning
Corning® BioCoat®
354688 354688
BD354688CS 466.15 USD InStock
BD354688 CACB354688
Corning® BioCoat® Poly-D-Lysine/Laminin 8-well Culture Slide, Corning
Microplates Cell Culture Plates
Corning® BioCoat™ 8-well culture slide with a uniform application of PDL/Laminin. Glass slide with polystyrene vessel, lid, and safety removal tool. 4/pack, 12/case. Corning BioCoat products are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures. Corning offers custom coating capabilities.
  • Shelf life: >= 3 months from date of shipment
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty