Would you like to log in again?
SP Bel-Art Stirrer Blades, Bel-Art Products, a part of SP
Supplier: Bel-Art
VWR®
F37971-0000 F37972-0000
58947-410CS 54.84 USD InStock
58947-410 58947-412
SP Bel-Art Stirrer Blades, Bel-Art Products, a part of SP
Stirrers Stirring Blades
Strong, non-scratch, inert, convenient
  • Inert and non-scratching
  • PTFE blade construction
  • Compatible with all standard button-tip stirring shafts
  • Autoclavable
“Long-groove” keyhole provides easy insertion into flask neck and proper position for stirring action.
Fits all standard button-tip stirring shafts
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty