Would you like to log in again?
Digital 'Pocket' Cutting Oil Refractometer, Model PAL-102S, ATAGO®
Supplier: Atago
4502
89008-814EA 806.03 USD InStock
89008-814
Digital 'Pocket' Cutting Oil Refractometer, Model PAL-102S, ATAGO®
Refractometers Handheld Refractometers
Designed for controlling the concentration of cutting oils
The device features IP65-rated dustproof and water-resistant protection, and Automatic Temperature Compensation for accurate measurements in shifting environmental conditions. A sample volume of 0.3mL is required for measurement results.
Molding lubricants and rust resistance liquids are also within the measurement range of this device. Refractometer is best suited for determining the percent dilution and controlling the percent concentration of hydraulic oil and cleaning liquids.
Ordering information: Uses two AAA batteries.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty