Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Starsys® Mobile Workstations, Metro™
Supplier: Metro International
Starsys®
SXRMWT36EBS4 SXRMWTK39EBS3 SXRMWD36EOH2 SXRMWT36EOH3 SXRMWT36EBS4 SXRMWTK39EBS3 SXRMWD36EOH2 SXRMWT36EBS6
98000-754EA 15735.49 USD InStock
98000-754 CA89237-256 89369-992 CA89369-990 89237-256 CA89237-276 89369-990 89237-276
Starsys® Mobile Workstations, Metro™
Furniture Casework
Workstation components are constructed of corrosion-proof polymer for a contamination-free product.
  • The countertops are 1" (25 mm) thick black epoxy resin
  • Overhead units include 27" (686 mm) cabinets with locking clear doors and two poly shelves
Microban® Antimicrobial product protection inhibits the growth of stain and odor-causing bacteria on Starsys, keeping the surface areas cleaner between cleanings. All preconfigured units come standard with taupe door/drawer pulls and polymer 5" (127 mm) Ø casters (2-total lock, 2-swivel).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty