Worktables feature advanced polymer construction that provides excellent corrosion resistance in the most corrosive lab environments.
- Tabletops feature smooth, easy-to-clean surfaces
- Constructed of either Type 304 stainless steel or Trespa® phenolic resin
- Mobile units are easily moved for cleaning
Lab worktables are load rated at 50 lbs. per square foot, up to a maximum of 600 lbs. when load is evenly distributed.
Ordering information: Casters (for mobile units) are sold separately.