Would you like to log in again?
Oil and Grease Wastewater Standard, NSI
Supplier: NSI Lab Solutions
QCI-069
NSQCI-069EA 79.74 USD InStock
NSQCI-069
Oil and Grease Wastewater Standard, NSI
Standards Environmental Analysis Standards
Ready-to-use whole volume standard for gravimetric analysis
Certified for total oil and grease in the range of 20–100mg/L. Includes CoA listing true value and acceptance limits. Supplied in glass bottle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty