Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
High-Level Solids Wastewater Standard, NSI
Supplier: NSI Lab Solutions
QCI-055H
NSQCI-055HCS 269.5 USD InStock
NSQCI-055H
High-Level Solids Wastewater Standard, NSI
Standards Environmental Analysis Standards
Certified for TDS, TS, and TSS
Standard contains 1000–3000 mg/L of TDS, 1400–3500 mg/L of TS, and 400–500 mg/L of TSS.
Includes CoA listing true value and acceptance limits.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty