Would you like to log in again?
EPA Method 8270 BNA Surrogates, NSI
Supplier: NSI Lab Solutions
WL-371-50
NSC-371-50EA 203.31 USD InStock
NSC-371-50
EPA Method 8270 BNA Surrogates, NSI
Standards Environmental Analysis Standards
Mixture is packaged in methanol at the stated concentrations in 50ml vial
Mixture contains 200µg/ml 2-Fluorophenol, 200µg/ml Phenol-d5, 200µg/ml 2,4,6-Tribromophenol, 100µg/ml 2-Fluorobiphenyl, 100µg/ml Nitrobenzene-d5, and 100µg/ml p-Terphenyl-d14.
An A2LA traceable standard that has been second source verified in house using an A2LA verified standard.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty