Would you like to log in again?
UST Diesel Range Organics (DRO) Standard (EPA/WISC), Restek
Supplier: Restek
31064
RK31064EA 62.08 USD InStock
RK31064
UST Diesel Range Organics (DRO) Standard (EPA/WISC), Restek
Standards Petroleum Industry Standards
Standards are packaged in methylene chloride at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains Decane (C10), Dodecane (C12), Tetradecane (C14), Hexadecane (C16), Octadecane (C18), Eicosane (C20), Docosane (C22), Tetracosane (C24), Hexacosane (C26), Octacosane (C28)
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty