Would you like to log in again?
Semi-Volatile Surrogate Standards for EPA methods, Restek
Supplier: Restek
31091
RK31091EA 32.98 USD InStock
RK31091
Semi-Volatile Surrogate Standards for EPA methods, Restek
Standards Environmental Analysis Standards
Standards are packaged in methylene chloride at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains 2-Fluorobiphenyl and 1-Fluoronaphthalene
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty