Would you like to log in again?
TCLP Base/Neutrals Standard, Restek
Supplier: Restek
31028
RK31028EA 40.74 USD InStock
RK31028
TCLP Base/Neutrals Standard, Restek
Standards Environmental Analysis Standards
Standard is packaged in acetone at a concentration of 2000 µg/mL in a 1 mL ampoule.
  • Mixture contains 1,4-Dichlorobenzene, 2,4-Dinitrotoluene, Hexachlorobenzene, Hexachlorobutadiene, Hexachloroethane, Nitrobenzene, and Pyridine
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty