Would you like to log in again?
UST BTEX Standard, Restek
Supplier: Restek
30051 30488
RK30051EA 51.41 USD InStock
RK30051 75834-404
UST BTEX Standard, Restek
Standards Petroleum Industry Standards
BTEX standards are packaged in purge-and-trap-grade methanol in a 1 mL ampoule.
Mixture contains 6 components; benzene, ethylbenzene, toluene, m-xylene, o-xylene, and p-xylene.
Delivery information: Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty