Would you like to log in again?
CLP–Pesticides Standard Mix B, Restek
Supplier: Restek
32004
RK32004EA 46.56 USD InStock
RK32004
CLP–Pesticides Standard Mix B, Restek
Standards Organic Standards
Standards are packaged in hexane at the stated concentrations in a 1 mL ampoule.
  • In acetone, 1 mL/ampoule
Each standard includes an ampoule breaker, deactivated amber screw-top vial, extra product label, Certificate of Analysis, and Material Safety Data Sheet (MSDS).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty