Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Nalgene® Lowboy™ Carboys with Spigot, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® Lowboy™
2324-0008 2324-0015
16117-977CS 1353.82 USD InStock
16117-977 16117-975
Nalgene® Lowboy™ Carboys with Spigot, Polypropylene, Thermo Scientific
Bottles Carboys
Carboys are ideal for storing and dispensing stock solutions.
  • Repeatedly autoclavable
  • Cap size: 63-415
  • Leakproof
Low-profile design fits easily in tight spaces, on shelves, or in refrigerator, and can be stacked three high. Integral handgrips for safe easy transport, and the polypropylene offers excellent chemical resistance.
Ordering information: Supplied with carboy, polypropylene spigot, and screw cap. For replacement caps, see cap 16219-104 in 16219-030 series.
Caution: Before autoclaving, set cap on top of carboy without engaging threads and disassemble spigot.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty