Would you like to log in again?
Tissue Culture Bottles, HiMedia
Supplier: HiMedia Laboratories
PW1125-50NO PW1125-50NO
95038-670CS 642.87 USD InStock
CA95038-670 95038-670
Tissue Culture Bottles, HiMedia
Bottles Cell Culture Bottles
Clear, autoclavable, polycarbonate jars with polypropylene lids.
Overflow capacity is 250 ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty