Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Citric acid, anhydrous ACS

Supplier: Columbus Chemical Industries, Inc
Warning
Synonyms: Hydroxytricarballylic acid

149000-57101
89088-068EA 526.22 USD InStock
89088-068
Citric acid, anhydrous ACS
Citric acid


Formula: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Boiling Pt: 310 °C (1013 hPa)
Melting Pt: 149…151 °C
Density: 1.665 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011669
CAS Number: 77-92-9
EINECS: 201-069-1
REACH: 01-2119457026-42-0000
Merck Index: 13,02350
https://us-legacy.vwr.com/stibo/web/std.lang.all/50/46/725046.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty