Would you like to log in again?
MicroGuard CE™ Hoods, International ENVIROGUARD™
Supplier: International Enviroguard
MicroGuard CE™
CE8063CI
89127-816CS 550.16 USD InStock
89127-816
MicroGuard CE™ Hoods, International ENVIROGUARD™
Disposable Garments
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting
Full face hoods are white with a tunnelized elastic face. Hood covers shoulders. Sizes are universal.
Tear-resistant. Recyclable.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty