Would you like to log in again?
Aerosol Dispenser, Veltek Associates
Supplier: Veltek
DEC-50
14003-314CS 1045.29 USD InStock
14003-314
Aerosol Dispenser, Veltek Associates
Soap Dispensers and Lotion Soap Dispensers
Hand-activated and hands-free dispensing mechanism eliminates cross-contamination from user to user during handling of the alcohol container.
The mechanism is constructed of 316L stainless steel and easily slides on or off a permanently installed wall plate that incorporates welded-pin connectors. Autoclavable.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty