Would you like to log in again?
VWR® Irradiated Beard Cover
Supplier: VWR
VWR®
414004-506
414004-506CS 125.66 USD InStock
414004-506
VWR® Irradiated Beard Cover
Disposable Garments
Beard cover is manufactured from strong, lightweight, and breathable spunbonded polypropylene (SPP) fabric, which protects against dirt, grime, and certain dry particulates in nonhazardous environments.
  • Strong, lightweight, breathable spunbonded polypropylene
  • Lowest level of particle shedding, non-linting
  • First-line barrier
  • Gamma irradiated to SAL of 10-4
To address the concern of bio-burden contamination, the beard cover is gamma irradiated to a Sterility Assurance Level (SAL) of 10-4. Manufactured and packaged in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.
Certifications: Beard cover is tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.
Ordering information: Packaged in sealed polyethylene bags, 10 per bag, contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty