Would you like to log in again?
Puritan® DNA-Free Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Puritan®
25-8061WCFDNA 25-8061WCFDNA
89131-648CS 222.04 USD InStock
89131-648 CA89131-648
Puritan® DNA-Free Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Swabs Swabs for Sampling
For crime scene investigation or specimen collection
Sturdy wood shaft. Absorbent cotton tip.
Certifications: Certified DNA-free and RNase-free.
Ordering information: Packaged 100 individually wrapped applicators per box, 10 boxes per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty