Would you like to log in again?
Fellowes® Jupiter™ JL 125 Laminator, Essendant
Supplier: Essendant
Fellowes® Jupiter™
FEL5734101
500016-350EA 564.31 USD InStock
500016-350
Fellowes® Jupiter™ JL 125 Laminator, Essendant
Laminators
High performance office laminator for a variety of professional applications.
  • Carrier-free 4-roller system for superior laminating results and consistent lamination quality
  • Quick heat-up time, ready in just four minutes
  • Fast and easy to use, finishes letter-size documents in about thirty seconds
  • Green light and audible beep signal when laminator is ready
  • Maximum document size is 12.5" wide
Includes laminating starter pack with ten 3-mil letter pouches. Versatile 4-roller laminator offers superior performance. Reverse mode disengages pouch for re-centering or removal. Unique clamshell design provides easy machine access for cleaning, maintenance, or misfeed removal. HeatGuard Technology™ keeps laminator cool to touch. Cold setting for laminating heat-sensitive materials in self-adhesive pouches.
Certifications: UL listed. cUL listed.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty