Would you like to log in again?
Clorox® Clean-Up® Cleaner with Bleach, Essendant
Supplier: Essendant
Clorox® Clean-Up®
CLO35420EA CLO35420CT CLO35417EA
500033-016EA 43.19 USD InStock
500033-016 500033-014 500032-410
Clorox® Clean-Up® Cleaner with Bleach, Essendant
Cleaning Agents Detergents and Surface Cleansers
Cleans, disinfects and deodorizes hard, nonporous surfaces in hospitals, nursing homes and institutional settings.
  • Removes tough stains such as blood and greasy dirt
Effective in 30 seconds against Pseudomonas, Salmonella, Staph, Rhinovirus, Hepatitis A and Influenza A2/Hong Kong virus.
Fragrance-free cleaner applicable on fiberglass, glass, plastic and stainless steel. Fresh fragrance cleaner used as general purpose cleaner.
Certifications: EPA Registered
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty