Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Cartridges, 113R00719, 113R00720, 113R00721, 113R00722, 113R00723, 113R00724, 113R00725, 113R00726, Essendant LLC MS

Supplier: Essendant
Xerox®
XER113R00726 XER113R00719 XER113R00725 XER113R00721 XER113R00723 XER113R00724 XER113R00720 XER113R00722
500028-169EA 248.71 USD InStock
500028-169 500028-170 500028-197 500028-196 500028-198 500028-168 500028-542 500028-167
Xerox® Cartridges, 113R00719, 113R00720, 113R00721, 113R00722, 113R00723, 113R00724, 113R00725, 113R00726, Essendant LLC MS
Inks
The advanced shading techniques of these cartridges makes ordinary presentations into extraordinary.

  • OEM and no remanufacturing available
  • Print at 5% page coverage
  • Cartridge type 6180 HC are providing high yield than the normal type
  • Installs quickly and easily, reducing downtime

Uses toner optimally to ensure that the prints look best.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty