Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xerox® Toner Cartridges, 106R01160, 106R01161, 106R01162, 106R01163, Essendant LLC MS

Supplier: Essendant
Xerox®
XER106R01163 XER106R01161 XER106R01160 XER106R01162
500029-471EA 815.54 USD InStock
500029-471 500029-470 500029-469 500029-468
Xerox® Toner Cartridges, 106R01160, 106R01161, 106R01162, 106R01163, Essendant LLC MS
Inks
These toners are designed to print high-quality documents and presentations with ease.

  • OEM and no remanufacturing available
  • Print at 5% page coverage
  • Reduces user interaction

These are produces sharper lines and crisper edges on every page.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty