Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Innovera® MICR Toner, 83640TMICR, Essendant LLC MS

Supplier: Essendant
Innovera®
IVR83640TMICR
500029-219EA 259.35 USD InStock
500029-219
Innovera® MICR Toner, 83640TMICR, Essendant LLC MS
Inks
Innovera® 83640T high-quality MICR toner product is uniquely formulated for peak performance.

  • Fully backed with a 100% performance guarantee
  • OEM and remanufacturing available
  • Cartridge with 5% coverage

Provides unrivalled printer compatibility and bank acceptance rates. Cartridge is easy to replace.

Certifications: Meets ANSI/ABA and all US and Canadian banking standards.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty