Would you like to log in again?
LYSOL® Disinfecting Wipes, Essendant
Supplier: Essendant
LYSOL®
RAC77182EA RAC81145 RAC77182CT
500032-704EA 12.57 USD InStock
500032-704 500037-840 500032-732
LYSOL® Disinfecting Wipes, Essendant
Wipes Disposable Wipes and Towels Disinfectant Wipes
Bleach- and alcohol-free disinfectant wipes clean and deodorize highly sensitive workspaces.
Safe on vinyl and nonporous surfaces, wipes provide maximum coverage on hospital beds, carts, counters and exam tables. Kills 99.9% of germs on hard, nonporous surfaces.
Certifications: EPA Registered.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty