Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lexmark™ Laser Cartridge, C930H2CG, C930H2KG, C930H2MG, Essendant LLC MS

Supplier: Essendant
Lexmark™
LEXC930H2KG LEXC930H2CG LEXC930H2MG
500028-715EA 1028.04 USD InStock
500028-715 500028-714 500037-406
Lexmark™ Laser Cartridge, C930H2CG, C930H2KG, C930H2MG, Essendant LLC MS
Inks
These laser cartridge creates crisp professional-quality prints and documents.

  • OEM and no remanufacturing available
  • Cartridge with 5% coverage

Expect fine lines, crisp edges, and sharp borders. High-capacity ensures maximum print volume.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty