Would you like to log in again?
Universal® Light-Duty Filament Tape, Essendant LLC MS
Supplier: Essendant
Universal®
UNV30018 UNV30048
500027-008RL 3.83 USD InStock
500027-008 500027-009
Universal® Light-Duty Filament Tape, Essendant LLC MS
Tapes
Packaging boxes and bundling doesn't always require the same strength tape--use this one for lighter, less industrial purposes
The general-purpose filament tape is ideal for non-critical packaging, unitizing and shipping. Tape features a film backing reinforced with fiberglass filament and has a 100 lbs. per inch tensile (bursting) strength. Comes with a 3" core.
Certifications: Meets U.S. Postal Regulations.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty