Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dataproducts® R7300 Typewriter Ribbon, Essendant

Supplier: Essendant
Dataproducts®
DPSR7300
500023-903EA 4.52 USD InStock
500023-903
Dataproducts® R7300 Typewriter Ribbon, Essendant
Typewriter Ribbons
For use with various typewriters.

  • Consistent, crisp character output
  • Typewriter device type
  • Singlestrike printing type
  • Compatible for use

Color: Black.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty