Would you like to log in again?
Easy-Paks® Neutral Cleaner
Supplier: Essendant
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
Easy Paks®
DVO990653EA
500032-796EA 45.17 USD InStock
500032-796
Easy-Paks® Neutral Cleaner
Cleaning Agents Detergents and Surface Cleansers
Bring back the shine to high-gloss floors, without having to worry about harming them in the process.
  • Cleaner powder leaves a fresh pine forest scent
Formula loosens and removes soil found in high-traffic areas, while protecting high-gloss, high solids finishes. Plus, the formula is phosphate-free and leaves no dulling film, without the need for rinsing. The premeasured packs of cleaning solution quickly dissolve in water, allowing you to start cleaning immediately.
Ideal for cleaning surfaces ranging from terrazzo, marble, slate, vinyl and vinyl asbestos, asphalt, ceramic and quarry tile.
Packaging: Packets come in a large, easy-to-store tub. 90 packets per tub.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty