Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Martin Yale® Model 1601 Ease-of-Use Tabletop AutoFolder™

Supplier: Martin Yale
Martin Yale® AutoFolder™
PRE1611
500044-040EA 1930.62 USD InStock
500044-040
Martin Yale® Model 1601 Ease-of-Use Tabletop AutoFolder™
Paper Folders and Joggers
Easy-to-use model ideal for mailrooms and offices with frequent medium to large size jobs.

  • High speed machine automatically feeds and folds a stack of documents up to 8.5×14"
  • Precision paper skew adjustment
  • Stapled sets of up to four sheets can be folded by hand-feeding the document into the feed table bypass
  • No fanning needed and the retarder is self-adjusting

Fold Types: Double Parallel, Half, Letter (for 8.5×11" sheets only), Right Angle, Z
9000 Sheets per Hour
Sheet Table Capacity: 150 Sheets
Convoyer, Automatic Jam Detection, Auto-Off
Manual Feed of Stapeled Sheets

Certifications: CE approved, CSA approved.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty