Would you like to log in again?
HON® 38000 Series Kneespace Credenza, Essendant LLC MS
Supplier: Essendant
HON®
HON38854NS HON38852NS HON38852CL HON38854CL
500006-980EA 1477.4 USD InStock
500006-980 500007-001 500043-036 500043-037
HON® 38000 Series Kneespace Credenza, Essendant LLC MS
Furniture Tables and Workbenches
Steel frame with steel-reinforced, high-pressure laminate top for superior rigidity and strength.
  • Drawers operate on heavy-duty steel ball bearing suspensions
  • Convenient cord management grommets
  • Radius edge style
  • Left: box/file, Right: box/file drawers
  • Double ³/₄ height, locking pedestal with 4 drawers
High-sided, full-extension file drawers accept hanging file folders; box drawers extend three-quarters. HON® 'One Key' core removable locks.
Certifications: Meets or exceeds ANSI/BIFMA standards.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty