Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safco® Boltless Steel Shelving

Supplier: Safco
Safco®
SAF5245BL SAF5246BL
500024-782EA 380.29 USD InStock
500024-782 500024-783
Safco® Boltless Steel Shelving
Furniture Shelves
Unique steel shelving requires no nuts or bolts. It assembles in minutes without tools.

  • Powder coated finish
  • Shelves adjust in 1" increments
  • Dent-resistant, rustproof, scratch resistant, washable

Versatile design allows two different set-ups: a five-shelf, 72" high storage unit, or a 36" high sturdy workbench.

Certifications: GREENGUARD Indoor Air Quality Certified®.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty