Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Filtrete™ Air Cleaning Replacement Filter, Essendant
Supplier: Essendant
Filtrete™
MMMOAC250RF MMMOAC150RF
500010-937EA 86.62 USD InStock
500010-937 500010-936
Filtrete™ Air Cleaning Replacement Filter, Essendant
Filters Humidifier Filters
Filtrete air cleaning filters snap easily into place after removing used filters.
Replace filter every three months to ensure maximum performance of you office air cleaner. Captures microscopic allergens like dust, smoke and smog particles, as well as large allergens like mold spores and pet dander from the air passing through the filter. Built-in activated carbon filter helps reduce odors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty