Would you like to log in again?
Fresh Products Super-Sorb Liquid Spills Absorbent, Essendant
Supplier: Essendant
Fresh Products
FRS614SSEA
500035-864EA 10.52 USD InStock
500035-864
Fresh Products Super-Sorb Liquid Spills Absorbent, Essendant
Spill Control Materials
Absorbs 60 times its weight immediately! Easy, sanitary disposal of blood, vomit, urine and other liquids from hard surfaces
Saves cleanup time—simply sprinkle granular absorbent onto spills, sweep up. Quat and microbiocide eliminate odors. Ideal for use in grocery stores, restaurants, hospitals, nursing homes, day care facilities, health clinics, etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty