Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alera® Essentia Series Swivel Task Chair

Supplier: Alera
Essentia
ALEVT48FA40B ALEVT48FA10B
500005-927EA 312.25 USD InStock
500005-927 500005-915
Alera® Essentia Series Swivel Task Chair
Chairs
Contoured seat and back provide added comfort for long hours at your desk.

  • Pneumatic seat height-adjustment
  • Seat depth adjustment
  • Durable 100% acrylic upholstery
  • Rotates a full 360° in either direction for ease of motion
  • Hard wheel tread

Black five-star base with five casters.

Optional adjustable T-arms ALE-VT49AKE10B available.

Certifications: GREENGUARD Indoor Air Quality Certified®, Meets or exceeds ANSI/BIFMA standards.

Ordering information: Optional arms sold and shipped separately. Casters supplied with this chair are not suitable for all floor types.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty